Please login to your online account to display your discounted pricing

5-Iodo-2-furancarboxaldehyde, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365840050

 View more versions of this product

Catalog No. AC365840050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder or Needles
Formula Weight 221.98
Color Light yellow to orange
Quantity 5g
Melting Point 124° to 129°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 5-Iodo-2-furancarboxaldehyde
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C5H3IO2
MDL Number MFCD00159503
CAS 2689-65-8
Synonym 5-Iodo-2-furaldehyde
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C
Assay Percent Range 97%

Specifications

Chemical Name or Material 5-Iodo-2-furancarboxaldehyde
CAS 2689-65-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H3IO2
Formula Weight 221.98
MDL Number MFCD00159503
Synonym 5-Iodo-2-furaldehyde
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Refrigerator +4°C

SDS & Safety Documents