Please login to your online account to display your discounted pricing

5-Iodo-2-furancarboxaldehyde, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Iodo-2-furancarboxaldehyde
CAS 2689-65-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H3IO2
Formula Weight 221.98
MDL Number MFCD00159503
Synonym 5-Iodo-2-furaldehyde
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC365840050

  SDS
thermo fisher scientific
AC365840050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Color Light yellow to orange
Physical Form Crystalline Powder or Needles
Formula Weight 221.98
Quantity 5g
Melting Point 124° to 129°C
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
CAS 2689-65-8
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C5H3IO2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Synonym 5-Iodo-2-furaldehyde
Packaging Glass bottle
Recommended Storage Refrigerator +4°C
Chemical Name or Material 5-Iodo-2-furancarboxaldehyde
MDL Number MFCD00159503
Assay Percent Range 97%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Refrigerator +4°C