missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-5-Iodo-2'-deoxyuridine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC122350250

 View more versions of this product

Catalog No. AC122350250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(+)-5-Iodo-2′-deoxyuridine
1% max. (1 g, 60°C) (vacuum)
+ 280.00 (20.00°C c=1,1M NaOH)
White to Beige
25g
98.5
99%
MFCD00134656
5-iodo-2'-deoxyuridine, 5-iododeoxyuridine, allergan 211, idoxene, idoxuridin, idoxuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin
XQFRJNBWHJMXHO-RRKCRQDMSA-N
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
5905
354.09
99%
Authentic
Glass bottle
+ 280.00
155.0°C to 180.0°C
54-42-2
100.0
C9H11IN2O5
15, 4931
Solubility in water: 1.6g/L (20°C). Other solubilities: 2.0g/L hcl 0.2n- 74.0g/L naoh 0.2n (25°C),4.4g/L methanol- 2.6g/L alcohol (25°C),0.014g/L ether- 0.003g/L chloroform (25°C),1.6g/L acetone- 1.8g/L ethyl acetate (25°C),5.7
C1C(C(OC1N2C=C(C(=O)NC2=O)I)CO)O
354.09
CHEBI:147675
Crystalline Powder

Chemical Identifiers

54-42-2
354.09
XQFRJNBWHJMXHO-RRKCRQDMSA-N
5905
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
C9H11IN2O5
MFCD00134656
5-iodo-2'-deoxyuridine, 5-iododeoxyuridine, allergan 211, idoxene, idoxuridin, idoxuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin
CHEBI:147675
C1C(C(OC1N2C=C(C(=O)NC2=O)I)CO)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Suspected of causing genetic defects.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging the unborn child.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-207-8

SDS Documents