Learn More
5-Iodo-2'-deoxycytidine, 99%, Thermo Scientific Chemicals
$351.03 - $671.64
Chemical Identifiers
CAS | 611-53-0 |
---|---|
Molecular Formula | C9H12IN3O4 |
Molecular Weight (g/mol) | 353.12 |
MDL Number | MFCD00038063 |
InChI Key | WEVJJMPVVFNAHZ-RRKCRQDMSA-N |
Synonym | 5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv |
PubChem CID | 65050 |
IUPAC Name | 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one |
SMILES | NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6135603
|
Thermo Scientific Chemicals
J6135603 |
1 g |
Each for $351.03
|
|
|||||
AAJ6135604
|
Thermo Scientific Chemicals
J6135604 |
2 g |
Each for $671.64
|
|
|||||
Description
5-Iodo-2'-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2?-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4,5:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Iodo-2′-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2′-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4′,5′:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Chemical Identifiers
611-53-0 | |
353.12 | |
WEVJJMPVVFNAHZ-RRKCRQDMSA-N | |
65050 | |
NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1 |
C9H12IN3O4 | |
MFCD00038063 | |
5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv | |
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one |
Specifications
611-53-0 | |
White | |
MFCD00038063 | |
5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv | |
WEVJJMPVVFNAHZ-RRKCRQDMSA-N | |
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one | |
65050 | |
99% | |
5-Iodo-2'-deoxycytidine |
180°C to 183°C | |
C9H12IN3O4 | |
1 g | |
Slightly soluble in water. | |
NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1 | |
353.12 | |
353.11 | |
Powder |
Safety and Handling
Harmful if swallowed
Acute toxicity (category 4)
EINECSNumber : 210-269-8
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only