Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Iodo-2,4-dimethoxypyrimidine, 98%

$71.12 - $708.07

Chemical Identifiers

Chemical Name or Material 5-Iodo-2,4-dimethoxypyrimidine
CAS 52522-99-3
Molecular Formula C6H7IN2O2
Formula Weight 266.04
MDL Number MFCD00090865
Synonym 2,4-Dimethoxy-5-iodopyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2496701

 
alfa aesar™
B2496701
0.25g Each for $71.12

AAB2496703

 
alfa aesar™
B2496703
1g Each for $203.19

AAB2496706

 
alfa aesar™
B2496706
5g Each for $708.07
Description & Specifications

Specifications

Formula Weight 266.04
Chemical Name or Material 5-Iodo-2,4-dimethoxypyrimidine
Assay 98%
Molecular Formula C6H7IN2O2
MDL Number MFCD00090865
CAS 52522-99-3
Melting Point 71° to 75°C
Synonym 2,4-Dimethoxy-5-iodopyrimidine
Quantity 0.25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.