missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Iodo-1,3-dimethyluracil, 99%, Thermo Scientific Chemicals

$121.38 - $359.55

Chemical Identifiers

CAS 40738-83-8
Molecular Formula C6H7IN2O2
Molecular Weight (g/mol) 266.04
MDL Number MFCD00192228
InChI Key XUUVRIXNFFFPCM-UHFFFAOYSA-N
Synonym 5-iodo-1,3-dimethyluracil, 5-iodo-1,3-dimethylpyrimidine-2,4 1h,3h-dione, 5-iodo-1,3-dimethyl-1h-pyrimidine-2,4-dione, acmc-209jgf, 1,3-dimethyl-5-iodouracil, 1,3-dimethyl 5-iodo uracil, uracil, 1,3-dimethyl-5-iodo, 2,4 1h,3h-pyrimidinedione,5-iodo-1,3-dimethyl, 5-iodo-1,3-dimethyl-1,3-dihydropyrimidine-2,4-dione
PubChem CID 622250
IUPAC Name 5-iodo-1,3-dimethylpyrimidine-2,4-dione
SMILES CN1C=C(I)C(=O)N(C)C1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2517303
View Documents Promotion Details
Thermo Scientific Chemicals
B2517303
1 g
Each for $121.38
 
AAB2517306
View Documents Promotion Details
Thermo Scientific Chemicals
B2517306
5 g
Each for $359.55
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40738-83-8
266.04
XUUVRIXNFFFPCM-UHFFFAOYSA-N
622250
CN1C=C(I)C(=O)N(C)C1=O
C6H7IN2O2
MFCD00192228
5-iodo-1,3-dimethyluracil, 5-iodo-1,3-dimethylpyrimidine-2,4 1h,3h-dione, 5-iodo-1,3-dimethyl-1h-pyrimidine-2,4-dione, acmc-209jgf, 1,3-dimethyl-5-iodouracil, 1,3-dimethyl 5-iodo uracil, uracil, 1,3-dimethyl-5-iodo, 2,4 1h,3h-pyrimidinedione,5-iodo-1,3-dimethyl, 5-iodo-1,3-dimethyl-1,3-dihydropyrimidine-2,4-dione
5-iodo-1,3-dimethylpyrimidine-2,4-dione

Specifications

40738-83-8
C6H7IN2O2
1 g
5-iodo-1,3-dimethyluracil, 5-iodo-1,3-dimethylpyrimidine-2,4 1h,3h-dione, 5-iodo-1,3-dimethyl-1h-pyrimidine-2,4-dione, acmc-209jgf, 1,3-dimethyl-5-iodouracil, 1,3-dimethyl 5-iodo uracil, uracil, 1,3-dimethyl-5-iodo, 2,4 1h,3h-pyrimidinedione,5-iodo-1,3-dimethyl, 5-iodo-1,3-dimethyl-1,3-dihydropyrimidine-2,4-dione
CN1C=C(I)C(=O)N(C)C1=O
266.04
266.04
5-Iodo-1,3-dimethyluracil
225°C (decomposition)
MFCD00192228
Light sensitive
XUUVRIXNFFFPCM-UHFFFAOYSA-N
5-iodo-1,3-dimethylpyrimidine-2,4-dione
622250
99%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only