Learn More
5-Indolylboronic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 359465000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Indolylboronic acid | |
96.0 | |
White | |
96% min. (HPLC) | |
C8H8BNO2 | |
500 mg | |
VHADYSUJZAPXOW-UHFFFAOYSA-N | |
1H-indol-5-ylboronic acid | |
2734361 | |
97% |
144104-59-6 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD01319013 | |
5-indolylboronic acid, indole-5-boronic acid, 1h-indol-5-yl boronic acid, 1h-indole-5-boronic acid, 5-indolboronic acid, 5-indolyl boronic acid, 5-indole-boronic acid, 1h-indol-5-ylboranediol, 5-indole boric acid, 5-indoleboronic acid | |
OB(O)C1=CC=C2NC=CC2=C1 | |
160.97 | |
160.96 | |
Crystalline Powder |
Chemical Identifiers
144104-59-6 | |
160.97 | |
VHADYSUJZAPXOW-UHFFFAOYSA-N | |
2734361 | |
OB(O)C1=CC=C2NC=CC2=C1 |
C8H8BNO2 | |
MFCD01319013 | |
5-indolylboronic acid, indole-5-boronic acid, 1h-indol-5-yl boronic acid, 1h-indole-5-boronic acid, 5-indolboronic acid, 5-indolyl boronic acid, 5-indole-boronic acid, 1h-indol-5-ylboranediol, 5-indole boric acid, 5-indoleboronic acid | |
1H-indol-5-ylboronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear pr
GHS Signal Word: Warning
RUO – Research Use Only