missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Hydroxypyridine-2-carboxylic Acid, TCI America™

$204.72 - $611.13

Chemical Identifiers

CAS 15069-92-8
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00661309
InChI Key CGRRUFNHHQCLDZ-UHFFFAOYSA-N
Synonym 5-hydroxypicolinic acid, 5-hydroxy-2-pyridinecarboxylic acid, 5-hydroxy-pyridine-2-carboxylic acid, 6-carboxy-3-pyridinol, 2-pyridinecarboxylic acid, 5-hydroxy, 5-hydroxy-2-pyridine carboxylic acid, pubchem10094, 5-hydroxypicolinic acidn, acmc-209d4x, ksc171o9d
PubChem CID 268740
IUPAC Name 5-hydroxypyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H14715G View Documents TCI America
H14715G
5g Each for $611.13
 
H14711G View Documents TCI America
H14711G
1g Each for $204.72
 
Specifications

Chemical Identifiers

15069-92-8
139.11
CGRRUFNHHQCLDZ-UHFFFAOYSA-N
268740
C1=CC(=NC=C1O)C(=O)O
C6H5NO3
MFCD00661309
5-hydroxypicolinic acid, 5-hydroxy-2-pyridinecarboxylic acid, 5-hydroxy-pyridine-2-carboxylic acid, 6-carboxy-3-pyridinol, 2-pyridinecarboxylic acid, 5-hydroxy, 5-hydroxy-2-pyridine carboxylic acid, pubchem10094, 5-hydroxypicolinic acidn, acmc-209d4x, ksc171o9d
5-hydroxypyridine-2-carboxylic acid

Specifications

White-Yellow
5g
C6H5NO3
5-hydroxypicolinic acid, 5-hydroxy-2-pyridinecarboxylic acid, 5-hydroxy-pyridine-2-carboxylic acid, 6-carboxy-3-pyridinol, 2-pyridinecarboxylic acid, 5-hydroxy, 5-hydroxy-2-pyridine carboxylic acid, pubchem10094, 5-hydroxypicolinic acidn, acmc-209d4x, ksc171o9d
C1=CC(=NC=C1O)C(=O)O
139.11
139.11
≥98.0% (HPLC,T)
268°C
15069-92-8
MFCD00661309
CGRRUFNHHQCLDZ-UHFFFAOYSA-N
5-hydroxypyridine-2-carboxylic acid
268740
Crystal-Powder at 20°C
5-Hydroxypyridine-2-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No