Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Hydroxypyridine-2-carboxylic acid, ≥98%

$110.61 - $110.61

Chemical Identifiers

Chemical Name or Material 5-Hydroxypyridine-2-carboxylic acid
CAS 15069-92-8
Formula Weight 139.11
MDL Number MFCD00661309
Beilstein 116009
Synonym 5-Hydroxypicolinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6426506

  SDS
alfa aesar™
H6426506
5g Each for $110.61

AAH6426514

  SDS
alfa aesar™
H6426514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6426503

  SDS
alfa aesar™
H6426503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.11
Melting Point 260° to 264°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-Hydroxypyridine-2-carboxylic acid
Physical Form Solid
Assay ≥98%
MDL Number MFCD00661309
CAS 15069-92-8
Beilstein 116009
Synonym 5-Hydroxypicolinic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g

Applications
Useful synthetic intermediate. Also intermediate for pharmaceutical application.

Solubility
Soluble in acetone, dimethyl sulfoxide , ethanol and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.