missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(Hydroxymethyl)uracil, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0168206
Description
5-(Hydroxymethyl)uracil serves as a stable major oxidative modification of thymine formed through the action of ionizing radiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-(Hydroxymethyl)uracil serves as a stable major oxidative modification of thymine formed through the action of ionizing radiation.
Solubility
Soluble in water. (50 g/L ) at 20°C, DMSO, dimethyl formamide or 100% ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
5-(Hydroxymethyl)uracil | |
>300°C | |
MFCD00056024 | |
125482 | |
Soluble in water. (50g/L ) at 20°C,DMSO,dimethyl formamide or 100% ethanol. | |
OCC1=CNC(=O)NC1=O | |
142.11 | |
CHEBI:16964 | |
98% |
4433-40-3 | |
C5H6N2O3 | |
5 g | |
5-hydroxymethyluracil, 5-hydroxymethyl uracil, 5-hydroxymethyl pyrimidine-2,4 1h,3h-dione, 5-oxymethyluracil, 4-methyl-5-oxyuracil, uracil, 5-hydroxymethyl, 2,4 1h,3h-pyrimidinedione, 5-hydroxymethyl, thymine, alpha-hydroxy, 5-hydroxymethyl-2,4 1h,3h-pyrimidinedione | |
JDBGXEHEIRGOBU-UHFFFAOYSA-N | |
5-(hydroxymethyl)-1H-pyrimidine-2,4-dione | |
78168 | |
142.12 |
Chemical Identifiers
4433-40-3 | |
142.11 | |
JDBGXEHEIRGOBU-UHFFFAOYSA-N | |
78168 | |
5-(hydroxymethyl)-1H-pyrimidine-2,4-dione |
C5H6N2O3 | |
MFCD00056024 | |
5-hydroxymethyluracil, 5-hydroxymethyl uracil, 5-hydroxymethyl pyrimidine-2,4 1h,3h-dione, 5-oxymethyluracil, 4-methyl-5-oxyuracil, uracil, 5-hydroxymethyl, 2,4 1h,3h-pyrimidinedione, 5-hydroxymethyl, thymine, alpha-hydroxy, 5-hydroxymethyl-2,4 1h,3h-pyrimidinedione | |
CHEBI:16964 | |
OCC1=CNC(=O)NC1=O |
Safety and Handling
EINECSNumber : 224-636-5
RTECSNumber : YR0513000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only