Learn More
5-Hydroxyisoquinoline, 90%, technical, Thermo Scientific™
Chemical Identifiers
CAS | 2439-04-5 |
---|---|
Molecular Formula | C9H7NO |
Molecular Weight (g/mol) | 145.16 |
MDL Number | MFCD00006906 |
InChI Key | CSNXUYRHPXGSJD-UHFFFAOYSA-N |
Synonym | 5-hydroxyisoquinoline, 5-isoquinolinol, 5-hydroxy-isoquinoline, chembl57481, 5-hydroxyisoquinolne, pubchem12826, acmc-2097ay, 5-hydroxy-2-azanaphthalene |
PubChem CID | 30386 |
IUPAC Name | isoquinolin-5-ol |
SMILES | C1=CC2=C(C=CN=C2)C(=C1)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC121290050
|
Thermo Scientific Chemicals
121290050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC121290250
|
Thermo Scientific Chemicals
121290250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2439-04-5 | |
145.16 | |
CSNXUYRHPXGSJD-UHFFFAOYSA-N | |
30386 | |
C1=CC2=C(C=CN=C2)C(=C1)O |
C9H7NO | |
MFCD00006906 | |
5-hydroxyisoquinoline, 5-isoquinolinol, 5-hydroxy-isoquinoline, chembl57481, 5-hydroxyisoquinolne, pubchem12826, acmc-2097ay, 5-hydroxy-2-azanaphthalene | |
isoquinolin-5-ol |
Specifications
2439-04-5 | |
100.0 | |
90% | |
C9H7NO | |
5g | |
5-hydroxyisoquinoline, 5-isoquinolinol, 5-hydroxy-isoquinoline, chembl57481, 5-hydroxyisoquinolne, pubchem12826, acmc-2097ay, 5-hydroxy-2-azanaphthalene | |
CSNXUYRHPXGSJD-UHFFFAOYSA-N | |
isoquinolin-5-ol | |
30386 | |
Granular Powder | |
Technical |
88.0 | |
Authentic | |
Glass bottle | |
MFCD00006906 | |
21,102 | |
Solubility in water: soluble | |
C1=CC2=C(C=CN=C2)C(=C1)O | |
145.16 | |
145.16 | |
90% | |
5-Hydroxyisoquinoline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
EINECSNumber : 219-456-9
RUO â Research Use Only