missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Hydroxyindole, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC121270050

 View more versions of this product

Catalog No. AC121270050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Hydroxyindole
Glass bottle
105.0°C to 110.0°C
1953-54-4
100.0
C8H7NO
21, III, 765
Solubility in water: insoluble in water. Other solubilities: soluble in acetone,ethanol and acetic acid
C1=CC2=C(C=CN2)C=C1O
133.15
133.15
97%
Authentic
Beige to Brown
5g
96.0
96% min. (GC)
MFCD00005677
5-hydroxy indole, 5-hydroxy-1h-indole, 5-hydroxy-indole, 5-hydroxyindol, 5-hydroxyindole, 5-indolol, hydroxy-5 indole, hydroxy-5-indole, indol-5-ol, unii-320un7xzyn
LMIQERWZRIFWNZ-UHFFFAOYSA-N
1H-indol-5-ol
16054
Crystalline Needles or Powder

Chemical Identifiers

1953-54-4
133.15
LMIQERWZRIFWNZ-UHFFFAOYSA-N
16054
C1=CC2=C(C=CN2)C=C1O
C8H7NO
MFCD00005677
5-hydroxy indole, 5-hydroxy-1h-indole, 5-hydroxy-indole, 5-hydroxyindol, 5-hydroxyindole, 5-indolol, hydroxy-5 indole, hydroxy-5-indole, indol-5-ol, unii-320un7xzyn
1H-indol-5-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-782-6

SDS Documents