missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Hydroxyindan 99.0+%, TCI America™
$144.33 - $437.97
Chemical Identifiers
CAS | 1470-94-6 |
---|---|
Molecular Formula | C9H10O |
Molecular Weight (g/mol) | 134.18 |
MDL Number | MFCD00003802 |
InChI Key | PEHSSTUGJUBZBI-UHFFFAOYSA-N |
Synonym | 5-indanol, indan-5-ol, 5-hydroxyindan, 5-hydroxyhydrindene, 5-hydroxyindane, 1h-inden-5-ol, 2,3-dihydro, indanol-5, unii-9z94h6f15t, 5-indano |
PubChem CID | 15118 |
ChEBI | CHEBI:59311 |
IUPAC Name | 2,3-dihydro-1H-inden-5-ol |
SMILES | OC1=CC=C2CCCC2=C1 |
Chemical Identifiers
1470-94-6 | |
134.18 | |
PEHSSTUGJUBZBI-UHFFFAOYSA-N | |
15118 | |
2,3-dihydro-1H-inden-5-ol |
C9H10O | |
MFCD00003802 | |
5-indanol, indan-5-ol, 5-hydroxyindan, 5-hydroxyhydrindene, 5-hydroxyindane, 1h-inden-5-ol, 2,3-dihydro, indanol-5, unii-9z94h6f15t, 5-indano | |
CHEBI:59311 | |
OC1=CC=C2CCCC2=C1 |
Specifications
1470-94-6 | |
White-Yellow | |
MFCD00003802 | |
2811 | |
PEHSSTUGJUBZBI-UHFFFAOYSA-N | |
2,3-dihydro-1H-inden-5-ol | |
15118 | |
134.18 | |
Crystalline Powder |
53°C | |
C9H10O | |
25 g | |
5-indanol, indan-5-ol, 5-hydroxyindan, 5-hydroxyhydrindene, 5-hydroxyindane, 1h-inden-5-ol, 2,3-dihydro, indanol-5, unii-9z94h6f15t, 5-indano | |
OC1=CC=C2CCCC2=C1 | |
134.18 | |
CHEBI:59311 | |
≥99.0% (GC) | |
5-Hydroxyindan |
Safety and Handling
RTECSNumber : NK7525000
TSCA : Yes