Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Hydroxy-2-(trifluoromethyl)pyridine, 97%

$106.36 - $293.82

Chemical Identifiers

Chemical Name or Material 5-Hydroxy-2-(trifluoromethyl)pyridine
CAS 216766-12-0
Molecular Formula C6H4F3NO
Formula Weight 163.1
MDL Number MFCD06801325
Synonym 6-Trifluoromethyl-3-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3173403

  SDS
alfa aesar™
H3173403
1g Each for $293.82

AAH31734MD

  SDS
alfa aesar™
H31734MD
250mg Each for $106.36
Description & Specifications

Specifications

Formula Weight 163.1
Melting Point 174° to 176°C
MDL Number MFCD06801325
Quantity 1g
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
Synonym 6-Trifluoromethyl-3-pyridinol
Health Hazard 1 Warning
Molecular Formula C6H4F3NO
CAS 216766-12-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 5-Hydroxy-2-(trifluoromethyl)pyridine
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.