missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Hydroxy-2-nitrobenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ L1205103

 View more versions of this product

Catalog No. AAL1205103



Description

Description

Applications
5-Hydroxy-2-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with air, oxidizing agents.
Specifications

Specifications

5-Hydroxy-2-nitrobenzaldehyde
1g
C7H5NO4
1910196
XLYPHUGUKGMURE-UHFFFAOYSA-N
5-hydroxy-2-nitrobenzaldehyde
39211
97%
166°C to 168°C
42454-06-8
MFCD00007332
3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
C1=CC(=C(C=C1O)C=O)[N+](=O)[O-]
167.12
167.12

Chemical Identifiers

42454-06-8
167.12
XLYPHUGUKGMURE-UHFFFAOYSA-N
39211
C1=CC(=C(C=C1O)C=O)[N+](=O)[O-]
C7H5NO4
MFCD00007332
3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
5-hydroxy-2-nitrobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CU6700000

Documents