Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Hydroxy-2-methylpyridine, 99%

$31.78 - $339.03

Chemical Identifiers

Chemical Name or Material 5-Hydroxy-2-methylpyridine
CAS 1121-78-4
Molecular Formula C6H7NO
Formula Weight 109.13
MDL Number MFCD00006339
Beilstein 107077
Synonym 5-Hydroxy-2-picoline; 6-Methyl-3-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2501406

  SDS
alfa aesar™
B2501406
5g Each for $31.78

AAB2501414

  SDS
alfa aesar™
B2501414
25g Each for $101.26

AAB2501422

  SDS
alfa aesar™
B2501422
100g Each for $339.03
Description & Specifications

Specifications

Formula Weight 109.13
Chemical Name or Material 5-Hydroxy-2-methylpyridine
Odor Odorless
Assay 99%
Molecular Formula C6H7NO
MDL Number MFCD00006339
CAS 1121-78-4
Beilstein 107077
Synonym 5-Hydroxy-2-picoline; 6-Methyl-3-pyridinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS Number UU7714900
Melting Point 167° to 171°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.