Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Hydroxy-2-methylindole, 98%

Chemical Identifiers

Chemical Name or Material 5-Hydroxy-2-methylindole
CAS 13314-85-7
Formula Weight 147.18
MDL Number MFCD00022714
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3558003

  SDS
alfa aesar™
H3558003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3558009

  SDS
alfa aesar™
H3558009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 130° to 132°C
Formula Weight 147.18
Storage Note 1 2° to 8°C
Color Brown
Assay 98%
CAS 13314-85-7
Quantity 1g
Physical Form Solid
Sensitivity Light Sensitive
Chemical Name or Material 5-Hydroxy-2-methylindole
MDL Number MFCD00022714
Safety and Handling

2° to 8°C