Learn More
5-Hydroxy-2-indolecarboxylic acid, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 162450010
Specifications
5-Hydroxy-2-indolecarboxylic acid | |
Glass bottle | |
249.0°C | |
21598-06-1 | |
100.0 | |
C9H7NO3 | |
5-hydroxy-1h-2-indolecarboxylic acid, 5-hydroxy-2-indolecarboxylic acid, 5-hydroxy-indole-2-carboxylic acid, 5-hydroxyindole-2-carboxylic acid, 5-hydroxyindole-2-carboxylicacid, acmc-20am33, indole-2-carboxylic acid, 5-hydroxy, pubchem1716, zlchem 754 | |
C1=CC2=C(C=C1O)C=C(N2)C(=O)O | |
177.159 | |
177.16 | |
98% |
Authentic | |
Colorless to Yellow | |
1g | |
97.5 | |
98% | |
MFCD00005615 | |
BIMHWDJKNOMNLD-UHFFFAOYSA-N | |
5-hydroxy-1H-indole-2-carboxylic acid | |
88958 | |
Fuming Liquid |
Chemical Identifiers
21598-06-1 | |
177.159 | |
BIMHWDJKNOMNLD-UHFFFAOYSA-N | |
88958 | |
C1=CC2=C(C=C1O)C=C(N2)C(=O)O |
C9H7NO3 | |
MFCD00005615 | |
5-hydroxy-1h-2-indolecarboxylic acid, 5-hydroxy-2-indolecarboxylic acid, 5-hydroxy-indole-2-carboxylic acid, 5-hydroxyindole-2-carboxylic acid, 5-hydroxyindole-2-carboxylicacid, acmc-20am33, indole-2-carboxylic acid, 5-hydroxy, pubchem1716, zlchem 754 | |
5-hydroxy-1H-indole-2-carboxylic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.