Please login to your online account to display your discounted pricing

5-Hydroxy-2-coumaranone, 97%, ACROS Organics™

$62.04 - $62.04

Chemical Identifiers

Chemical Name or Material 5-Hydroxy-2-coumaranone
CAS 2688-48-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H6O3
Formula Weight 150.13
MDL Number MFCD00005857
Synonym 2, 5-Dihydroxyphenylacetic acid γ-lactone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC173380010

  SDS
thermo fisher scientific
173380010
Glass bottle 1g Each for $62.04
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Fine Crystalline Powder
Formula Weight 150.13
Quantity 1g
Color Beige
Melting Point 190° to 195°C
Beilstein 18, 17
CAS Max % 100.0
CAS Min % 96.0
Synonym 2, 5-Dihydroxyphenylacetic acid γ-lactone
Assay Percent Range 97%
Chemical Name or Material 5-Hydroxy-2-coumaranone
Molecular Formula C8H6O3
CAS 2688-48-4
MDL Number MFCD00005857
Packaging Glass bottle
Infrared Spectrum Authentic