Learn More
5-Hydroxy-1,4-naphthoquinone, 99%, Thermo Scientific Chemicals
$96.04 - $282.85
Chemical Identifiers
CAS | 481-39-0 |
---|---|
Molecular Formula | C10H6O3 |
Molecular Weight (g/mol) | 174.16 |
MDL Number | MFCD00001684 |
InChI Key | KQPYUDDGWXQXHS-UHFFFAOYSA-N |
Synonym | juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon |
PubChem CID | 3806 |
ChEBI | CHEBI:15794 |
IUPAC Name | 5-hydroxynaphthalene-1,4-dione |
SMILES | OC1=CC=CC2=C1C(=O)C=CC2=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2834303
|
Thermo Scientific Chemicals
H2834303 |
1 g |
Each for $96.04
|
|
|||||
AAH2834306
|
Thermo Scientific Chemicals
H2834306 |
5 g |
Each for $282.85
|
|
|||||
Description
5-Hydroxy-1,4-naphthoquinone is used as a natural dye in cloth, fabrics, wool and ink. It is a coloring agent for food and cosmetics. It is involved in the synthesis of poly(hydroxyl-1,4-naphthoquinone) stabilized gold nanoparticles (AuNQ NPs), which is used for nonenzymatic electrochemical detection of glucose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Hydroxy-1,4-naphthoquinone is used as a natural dye in cloth, fabrics, wool and ink. It is a coloring agent for food and cosmetics. It is involved in the synthesis of poly(hydroxyl-1,4-naphthoquinone) stabilized gold nanoparticles (AuNQ NPs), which is used for nonenzymatic electrochemical detection of glucose.
Solubility
Soluble in alcohol, chloroform, dimethylsulfoxide, ether and aqueous solution of alkalies. Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong bases, strong reducing agents and strong oxidizing agents.
Chemical Identifiers
481-39-0 | |
174.16 | |
KQPYUDDGWXQXHS-UHFFFAOYSA-N | |
3806 | |
5-hydroxynaphthalene-1,4-dione |
C10H6O3 | |
MFCD00001684 | |
juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon | |
CHEBI:15794 | |
OC1=CC=CC2=C1C(=O)C=CC2=O |
Specifications
481-39-0 | |
C10H6O3 | |
1 g | |
Light sensitive | |
juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon | |
KQPYUDDGWXQXHS-UHFFFAOYSA-N | |
5-hydroxynaphthalene-1,4-dione | |
3806 | |
174.16 | |
5-Hydroxy-1,4-naphthoquinone |
161°C to 163°C | |
MFCD00001684 | |
UN2811 | |
14,5629 | |
Soluble in alcohol,chloroform,dimethylsulfoxide,ether and aqueous solution of alkalies. Slightly soluble in water. | |
OC1=CC=CC2=C1C(=O)C=CC2=O | |
174.16 | |
CHEBI:15794 | |
99% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 207-567-5
RTECSNumber : QJ5775000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only