Learn More
5-Hexynoic acid, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2513214
Description
5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
5-Hexynoic acid | |
0.99 | |
>110°C (230°F) | |
1.4495 | |
25 g | |
1743192 | |
Miscible with water. | |
C#CCCCC(=O)O | |
112.128 | |
CHEBI:73511 | |
97% |
53293-00-8 | |
73°C to 74°C (0.2 mmHg) | |
C6H8O2 | |
MFCD00066346 | |
UN3265 | |
5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h | |
VPFMEXRVUOPYRG-UHFFFAOYSA-N | |
hex-5-ynoic acid | |
143036 | |
112.13 |
Chemical Identifiers
53293-00-8 | |
112.128 | |
VPFMEXRVUOPYRG-UHFFFAOYSA-N | |
143036 | |
hex-5-ynoic acid |
C6H8O2 | |
MFCD00066346 | |
5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h | |
CHEBI:73511 | |
C#CCCCC(=O)O |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only