Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Hexyn-3-ol, 98+%

Chemical Identifiers

Chemical Name or Material 5-Hexyn-3-ol
CAS 19780-84-8
Molecular Formula C6H10O
Formula Weight 98.15
MDL Number MFCD00041593
UN Number UN1987
Beilstein 1698904
Synonym Ethyl propargyl carbinol; 1-Hexyn-4-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0543703

 
alfa aesar™
L0543703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0543706

 
alfa aesar™
L0543706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.15
Chemical Name or Material 5-Hexyn-3-ol
Assay ≥98%
Molecular Formula C6H10O
MDL Number MFCD00041593
Synonym Ethyl propargyl carbinol; 1-Hexyn-4-ol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
CAS 19780-84-8
UN Number UN1987
Beilstein 1698904
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Flash Point 43°C (109°F)
Boiling Point 58° to 59°C (25mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.