Please login to your online account to display your discounted pricing

5-Hexyn-1-ol, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC296580050

 View more versions of this product

Catalog No. AC296580050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (GC)
Physical Form Liquid
Formula Weight 98.14
Quantity 5g
Color Clear colorless to yellow
Density 0.89g/mL
Chemical Name or Material 5-Hexyn-1-ol
Assay Percent Range 97%
Linear Formula HC~2C(CH2)4OH
Infrared Spectrum Authentic
Molecular Formula C6H10O
Packaging Glass bottle
Refractive Index 1.4470 to 1.4510 (20°C, 589nm)
CAS 928-90-5

Specifications

Chemical Name or Material 5-Hexyn-1-ol
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C6H10O
Linear Formula HC~2C(CH2)4OH
Formula Weight 98.14