missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Hexenyl acetate, 97%

Manufacturer:  Alfa Aesar™ H5344522

 View more versions of this product

Catalog No. AAH5344522



Description

Description

Applications
(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.

Solubility
Insoluble in water, soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

5-Hexenyl acetate
60°C (142°F)
173°C to 174°C
5048-26-0
CH3CO2(CH2)4CH=CH2
UN3272
Insoluble in water,soluble in alcohol.
CC(=O)OCCCCC=C
142.198
142.2
0.883
1.423
100g
C8H14O2
MFCD00060856
5-hexen-1-ol, acetate, 5-hexene-1-ol, acetate, 5-hexenyl acetate, 5-hexenyl acetate #, 6-acetoxy-1-hexene, 6-acetoxyhex-1-ene, acetic acid 5-hexen-1-yl ester, acetic acid 5-hexenyl ester, acmc-1aw0j, hex-5-en-1-yl acetate
MPLWNENKBSBMFN-UHFFFAOYSA-N
hex-5-enyl acetate
537536
97%

Chemical Identifiers

5048-26-0
142.198
MPLWNENKBSBMFN-UHFFFAOYSA-N
537536
CC(=O)OCCCCC=C
C8H14O2
MFCD00060856
5-hexen-1-ol, acetate, 5-hexene-1-ol, acetate, 5-hexenyl acetate, 5-hexenyl acetate #, 6-acetoxy-1-hexene, 6-acetoxyhex-1-ene, acetic acid 5-hexen-1-yl ester, acetic acid 5-hexenyl ester, acmc-1aw0j, hex-5-en-1-yl acetate
hex-5-enyl acetate
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents