missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Hexenenitrile, TCI America™

$206.25 - $708.91

Chemical Identifiers

CAS 5048-19-1
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.145
MDL Number MFCD00067056
InChI Key UNAQSRLBVVDYGP-UHFFFAOYSA-N
Synonym 5-hexenenitrile, 5-cyano-1-pentene, 4-pentenyl cyanide, hex-1-ene-6-nitrile, acmc-209kmu, 5-hexenenitrile
PubChem CID 123222
IUPAC Name hex-5-enenitrile
SMILES C=CCCCC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H083625ML View Documents TCI America
H083625ML
25mL Each for $708.91
 
H08365ML View Documents TCI America
H08365ML
5mL Each for $206.25
 
Specifications

Chemical Identifiers

5048-19-1
95.145
UNAQSRLBVVDYGP-UHFFFAOYSA-N
123222
C=CCCCC#N
C6H9N
MFCD00067056
5-hexenenitrile, 5-cyano-1-pentene, 4-pentenyl cyanide, hex-1-ene-6-nitrile, acmc-209kmu, 5-hexenenitrile
hex-5-enenitrile

Specifications

162°C
25mL
C6H9N
1992
UNAQSRLBVVDYGP-UHFFFAOYSA-N
hex-5-enenitrile
123222
Clear Liquid at 20°C
5-Hexenenitrile
Yellow
5048-19-1
MFCD00067056
5-hexenenitrile, 5-cyano-1-pentene, 4-pentenyl cyanide, hex-1-ene-6-nitrile, acmc-209kmu, 5-hexenenitrile
C=CCCCC#N
95.145
95.15
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No