Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Hexenenitrile, 97%

$66.34 - $66.34

Chemical Identifiers

Chemical Name or Material 5-Hexenenitrile
CAS 5048-19-1
Molecular Formula C6H9N
Formula Weight 95.15
MDL Number MFCD00067056
UN Number UN1992
Beilstein 1739773
Synonym 5-Cyano-1-pentene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1096803

 
alfa aesar™
L1096803*
1g Each for $66.34

AAL1096806

 
alfa aesar™
L1096806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 95.15
Assay 97%
Odor Recognizable
Chemical Name or Material 5-Hexenenitrile
Molecular Formula C6H9N
MDL Number MFCD00067056
CAS 5048-19-1
UN Number UN1992
Beilstein 1739773
Synonym 5-Cyano-1-pentene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Flash Point 40°C (104°F)
Boiling Point 160° to 162°C
Quantity 1g

Applications
5-Hexenenitrile, is used as a natural substances and extractives.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.