missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Hexenenitrile, 95%, ACROS Organics™

$240.33 - $240.33

Chemical Identifiers

CAS 5048-19-1
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD00067056
InChI Key UNAQSRLBVVDYGP-UHFFFAOYSA-N
Synonym 4-pentenyl cyanide, 5-cyano-1-pentene, 5-hexenenitrile, acmc-209kmu, hex-1-ene-6-nitrile
PubChem CID 123222
IUPAC Name hex-5-enenitrile
SMILES C=CCCCC#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC259810010
SDS View Documents
Thermo Fisher Scientific
AC259810010
1mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC259810050
SDS View Documents
Thermo Fisher Scientific
259810050
5mL Glass bottle
Each for $240.33
 
AC259810250
SDS View Documents
Thermo Fisher Scientific
AC259810250
25mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

5048-19-1
95.15
UNAQSRLBVVDYGP-UHFFFAOYSA-N
123222
C=CCCCC#N
C6H9N
MFCD00067056
4-pentenyl cyanide, 5-cyano-1-pentene, 5-hexenenitrile, acmc-209kmu, hex-1-ene-6-nitrile
hex-5-enenitrile

Specifications

0.8200g/mL
Glass bottle
0.82
1mL
94.0
94% min. (GC)
CH2=CHCH2CH2CH2CN
4-pentenyl cyanide, 5-cyano-1-pentene, 5-hexenenitrile, acmc-209kmu, hex-1-ene-6-nitrile
UNAQSRLBVVDYGP-UHFFFAOYSA-N
hex-5-enenitrile
123222
95%
40°C
1.4250 to 1.4270
162.0°C to 163.0°C
5048-19-1
100.0
C6H9N
MFCD00067056
Solubility in water: soluble in hot water.
C=CCCCC#N
95.15
95.15
5-Hexenenitrile, 95%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapour.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove

GHS Signal Word: Warning

SDS