missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Hexenenitrile, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 259810050

 View more versions of this product

Catalog No. AC259810050



Description

Description

Specifications

Specifications

5-Hexenenitrile
40°C
1.4250 to 1.4270
162.0°C to 163.0°C
95%
94.0
94% min. (GC)
CH2=CHCH2CH2CH2CN
4-pentenyl cyanide, 5-cyano-1-pentene, 5-hexenenitrile, acmc-209kmu, hex-1-ene-6-nitrile
UNAQSRLBVVDYGP-UHFFFAOYSA-N
hex-5-enenitrile
123222
95%
0.8200g/mL
Glass bottle
0.82
5mL
5048-19-1
100.0
C6H9N
MFCD00067056
Solubility in water: soluble in hot water.
C=CCCCC#N
95.15
95.15

Chemical Identifiers

5048-19-1
95.15
UNAQSRLBVVDYGP-UHFFFAOYSA-N
123222
C=CCCCC#N
C6H9N
MFCD00067056
4-pentenyl cyanide, 5-cyano-1-pentene, 5-hexenenitrile, acmc-209kmu, hex-1-ene-6-nitrile
hex-5-enenitrile
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapour.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove

GHS Signal Word: Warning

SDS Documents