missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Hexen-2-one, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC120781000
Specifications
5-Hexen-2-one | |
23°C | |
1.4190 to 1.421 | |
128°C to 129°C | |
109-49-9 | |
100.0 | |
C6H10O | |
MFCD00008793 | |
1-hexen-5-one, 1-hexene-5-one, 3-buten-1-yl methyl ketone, 5-hexen-2-one, 5-hexene-2-one, 5-oxo-1-hexene, allyl acetone, allylacetone, unii-xwm6rl9yei, xwm6rl9yei | |
RNDVGJZUHCKENF-UHFFFAOYSA-N | |
hex-5-en-2-one | |
7989 | |
98% |
0.8400g/mL | |
Glass bottle | |
0.84 | |
100mL | |
97.5 | |
97.5% min. (GC) | |
H2C=CHCH2CH2COCH3 | |
01, 734 | |
Solubility in water: soluble in water. Other solubilities: soluble in organic solvents | |
CC(=O)CCC=C | |
98.14 | |
98.14 |
Chemical Identifiers
109-49-9 | |
98.14 | |
RNDVGJZUHCKENF-UHFFFAOYSA-N | |
7989 | |
CC(=O)CCC=C |
C6H10O | |
MFCD00008793 | |
1-hexen-5-one, 1-hexene-5-one, 3-buten-1-yl methyl ketone, 5-hexen-2-one, 5-hexene-2-one, 5-oxo-1-hexene, allyl acetone, allylacetone, unii-xwm6rl9yei, xwm6rl9yei | |
hex-5-en-2-one |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-675-1