Learn More
5-Hexen-2-one, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2250514
Description
Allyl Acetone is used as a nature identical flavor and fragrance. And it is also used to produce 2-methyl-hex-5-en-2-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAllyl Acetone is used as a nature identical flavor and fragrance. And it is also used to produce 2-methyl-hex-5-en-2-ol.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
5-Hexen-2-one | |
0.847 | |
23°C (73°F) | |
C6H10O | |
MFCD00008793 | |
UN1224 | |
5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone | |
RNDVGJZUHCKENF-UHFFFAOYSA-N | |
hex-5-en-2-one | |
7989 | |
≥98% |
109-49-9 | |
127°C to 128°C | |
Like Ketone | |
1.42 | |
25 g | |
1699137 | |
Soluble in water. | |
CC(=O)CCC=C | |
98.145 | |
98.15 |
Chemical Identifiers
109-49-9 | |
98.145 | |
RNDVGJZUHCKENF-UHFFFAOYSA-N | |
7989 | |
CC(=O)CCC=C |
C6H10O | |
MFCD00008793 | |
5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone | |
hex-5-en-2-one |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber : 203-675-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only