Learn More
5-Hexen-1-ol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1576606
Description
5-Hexen-1-ol is used in cyclization to a tetrahydropyran by phenylselenoetherification. It is also used as a building block in synthetic chemistry. Further, it is used to prepare 6-bromo-hex-1-ene by reaction with phosphorus tribromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Hexen-1-ol is used in cyclization to a tetrahydropyran by phenylselenoetherification. It is also used as a building block in synthetic chemistry. Further, it is used to prepare 6-bromo-hex-1-ene by reaction with phosphorus tribromide.
Solubility
Miscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
Specifications
5-Hexen-1-ol | |
<°C to 20°C | |
152°C to 155°C | |
97% | |
1.436 | |
5 g | |
1236458 | |
Miscible with water. | |
C=CCCCCO | |
100.161 | |
100.16 | |
Liquid |
821-41-0 | |
0.842 | |
59°C (138°F) | |
C6H12O | |
MFCD00002981 | |
UN2924 | |
5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2 | |
UIZVMOZAXAMASY-UHFFFAOYSA-N | |
hex-5-en-1-ol | |
69963 | |
98% |
Chemical Identifiers
821-41-0 | |
100.161 | |
UIZVMOZAXAMASY-UHFFFAOYSA-N | |
69963 | |
C=CCCCCO |
C6H12O | |
MFCD00002981 | |
5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2 | |
hex-5-en-1-ol |
Safety and Handling
GHS H Statement
H314-H318-H226
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 212-477-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only