Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Hexadecanone, 99%

$67.42 - $748.09

Chemical Identifiers

Chemical Name or Material 5-Hexadecanone
CAS 41903-81-5
Molecular Formula C16H32O
Formula Weight 240.43
MDL Number MFCD00051550
Synonym n-Butyl n-undecyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2488106

 
alfa aesar™
B2488106
5g Each for $67.42

AAB2488114

 
alfa aesar™
B2488114
25g Each for $238.59

AAB2488122

 
alfa aesar™
B2488122
100g Each for $748.09
Description & Specifications

Specifications

Formula Weight 240.43
Chemical Name or Material 5-Hexadecanone
Assay 99%
Molecular Formula C16H32O
MDL Number MFCD00051550
CAS 41903-81-5
Synonym n-Butyl n-undecyl ketone
Melting Point 36° to 38°C
Quantity 5g