Please login to your online account to display your discounted pricing

5-Hex-1-ynylnicotinic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Hex-1-ynylnicotinic acid
CAS 306935-31-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H13NO2
Formula Weight 203.24
MDL Number MFCD00830967
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM09434DA

 
fisher scientific company
KM09434DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 203.24
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 5-Hex-1-ynylnicotinic acid
Assay Percent Range 97%
Molecular Formula C12H13NO2
Packaging Amber glass bottle
MDL Number MFCD00830967
CAS 306935-31-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.