Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Formyl-4-methylthiophene-2-boronic acid 1,3-propanediol ester, 95%

$131.30 - $131.30

Chemical Identifiers

Chemical Name or Material 5-Formyl-4-methylthiophene-2-boronic acid 1,3-propanediol ester
CAS 374537-98-1
Molecular Formula C9H11BO3S
Formula Weight 210.06g/mol
MDL Number MFCD02093958
Synonym 5-(1,3,2-Dioxaborinan-2-yl)-3-methylthiophene-2-carboxaldehyde; 2-(2-Formyl-3-methylthien-5-yl)-1,3,2-dioxaborinane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1777903

 
alfa aesar™
L17779-03
1g Each for $131.30
Description & Specifications

Specifications

Melting Point ≈66°C
Quantity 1g
Chemical Name or Material 5-Formyl-4-methylthiophene-2-boronic acid 1,3-propanediol ester
CAS 374537-98-1
Assay 95%
Molecular Formula C9H11BO3S
Formula Weight 210.06g/mol
MDL Number MFCD02093958
Synonym 5-(1,3,2-Dioxaborinan-2-yl)-3-methylthiophene-2-carboxaldehyde; 2-(2-Formyl-3-methylthien-5-yl)-1,3,2-dioxaborinane