Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Formyl-2-methoxybenzeneboronic acid, 98%

$73.69 - $195.88

Chemical Identifiers

Chemical Name or Material 5-Formyl-2-methoxybenzeneboronic acid
CAS 127972-02-5
Molecular Formula C8H9BO4
Formula Weight 179.97
MDL Number MFCD01319044
Beilstein 5334095
Synonym 3-Borono-p-anisaldehyde; 3-Borono-4-methoxybenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1905903

  SDS
alfa aesar™
L1905903
1g Each for $195.88

AAL19059MD

  SDS
alfa aesar™
L19059MD
250mg Each for $73.69
Description & Specifications

Specifications

Formula Weight 179.97
Chemical Name or Material 5-Formyl-2-methoxybenzeneboronic acid
CAS 127972-02-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Assay 98%
MDL Number MFCD01319044
Molecular Formula C8H9BO4
Synonym 3-Borono-p-anisaldehyde; 3-Borono-4-methoxybenzaldehyde
Beilstein 5334095
Melting Point ≈160°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.