Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Formamido-1-(2-formyloxyethyl)-1H-pyrazole, 97%

Chemical Identifiers

Chemical Name or Material 5-Formamido-1-(2-formyloxyethyl)-1H-pyrazole
CAS 116856-18-9
Formula Weight 183.17
MDL Number MFCD09744040
Beilstein 7762121
Synonym 2-(5-Formamido-1H-pyrazol-1-yl)ethyl formate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6670103

 
alfa aesar™
H6670103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6670106

 
alfa aesar™
H6670106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.17
Chemical Name or Material 5-Formamido-1-(2-formyloxyethyl)-1H-pyrazole
CAS 116856-18-9
Physical Form Solid
MDL Number MFCD09744040
Assay 97%
Beilstein 7762121
Synonym 2-(5-Formamido-1H-pyrazol-1-yl)ethyl formate
Quantity 1g