Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoropyridine-2-carboxylic acid, 97%

$139.16 - $386.36

Chemical Identifiers

Chemical Name or Material 5-Fluoropyridine-2-carboxylic acid
CAS 107504-08-5
Molecular Formula C6H4FNO2
Formula Weight 141.1
MDL Number MFCD04114196
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3208603

  SDS
alfa aesar™
H3208603
1g Each for $386.36

AAH32086MD

  SDS
alfa aesar™
H32086MD
250mg Each for $139.16
Description & Specifications

Specifications

Formula Weight 141.1
Quantity 1g
Chemical Name or Material 5-Fluoropyridine-2-carboxylic acid
Assay 97%
Molecular Formula C6H4FNO2
MDL Number MFCD04114196
CAS 107504-08-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 164° to 166°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.