missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Fluoroorotic Acid Monohydrate 95.0+%, TCI America™
$26.54 - $81.29
Chemical Identifiers
CAS | 220141-70-8 |
---|---|
Molecular Formula | C5H5FN2O5 |
Molecular Weight (g/mol) | 192.10 |
MDL Number | MFCD00150658 |
InChI Key | LODRRYMGPWQCTR-UHFFFAOYSA-N |
Synonym | 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, 5-fluoroorotic acid monohydrate, 5-fluoroorotic acid hydrate, 5-fluoroorotic acid,monohydrate, 5-fluoro-1,2,3,6-tetrahydro-2,6-dioxopyrimidine-4-carboxylic acid hydrate, 5-fluoroortic acid monohydrate, 5-fluorouracil-4-carboxylic acid hydrate, 5-fluoroorotic acid n-hydrate, 5-fluororotic acid monohydrate, 5-fluoro-orotic acid monohydrate |
PubChem CID | 16212749 |
IUPAC Name | 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate |
SMILES | O.OC(=O)C1=C(F)C(=O)NC(=O)N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
F0382100MG
|
TCI America
F0382100MG |
100 mg |
Each for $26.54
|
|
|||||
F03821G
|
TCI America
F03821G |
1 g |
Each for $81.29
|
|
|||||
Chemical Identifiers
220141-70-8 | |
192.10 | |
LODRRYMGPWQCTR-UHFFFAOYSA-N | |
16212749 | |
O.OC(=O)C1=C(F)C(=O)NC(=O)N1 |
C5H5FN2O5 | |
MFCD00150658 | |
5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, 5-fluoroorotic acid monohydrate, 5-fluoroorotic acid hydrate, 5-fluoroorotic acid,monohydrate, 5-fluoro-1,2,3,6-tetrahydro-2,6-dioxopyrimidine-4-carboxylic acid hydrate, 5-fluoroortic acid monohydrate, 5-fluorouracil-4-carboxylic acid hydrate, 5-fluoroorotic acid n-hydrate, 5-fluororotic acid monohydrate, 5-fluoro-orotic acid monohydrate | |
5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate |
Specifications
220141-70-8 | |
MFCD00150658 | |
5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, 5-fluoroorotic acid monohydrate, 5-fluoroorotic acid hydrate, 5-fluoroorotic acid,monohydrate, 5-fluoro-1,2,3,6-tetrahydro-2,6-dioxopyrimidine-4-carboxylic acid hydrate, 5-fluoroortic acid monohydrate, 5-fluorouracil-4-carboxylic acid hydrate, 5-fluoroorotic acid n-hydrate, 5-fluororotic acid monohydrate, 5-fluoro-orotic acid monohydrate | |
O.OC(=O)C1=C(F)C(=O)NC(=O)N1 | |
192.10 | |
174.09 | |
Crystalline Powder |
C5H5FN2O5 | |
100 mg | |
LODRRYMGPWQCTR-UHFFFAOYSA-N | |
5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate | |
16212749 | |
≥95.0% (HPLC,T) | |
5-Fluoroorotic Acid Monohydrate |
Safety and Handling
RTECSNumber : UV7800000
TSCA : No
Recommended Storage : Refrigerator