missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Fluoronicotinic acid, 98%, Thermo Scientific Chemicals
$63.58 - $673.25
Chemical Identifiers
CAS | 402-66-4 |
---|---|
Molecular Formula | C6H4FNO2 |
Molecular Weight (g/mol) | 141.10 |
MDL Number | MFCD01318537 |
InChI Key | BXZSBDDOYIWMGC-UHFFFAOYSA-N |
Synonym | 5-fluoronicotinic acid, 5-fluoro-3-pyridinecarboxylic acid, 5-fluoro-nicotinic acid, 3-pyridinecarboxylic acid, 5-fluoro, nicotinic acid, 5-fluoro, 3-fluoro-5-carboxypyridine, 3-fluoro-5-pyridinecarboxylic acid, 5-fluoronicotinicacid, pubchem10942 |
PubChem CID | 120232 |
IUPAC Name | 5-fluoropyridine-3-carboxylic acid |
SMILES | OC(=O)C1=CN=CC(F)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6116206
|
Thermo Scientific Chemicals
H6116206 |
5 g |
Each for $63.58
|
|
|||||
AAH6116214
|
Thermo Scientific Chemicals
H6116214 |
25 g |
Each for $238.62
|
|
|||||
AAH6116222
|
Thermo Scientific Chemicals
H6116222 |
100 g |
Each for $673.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Fluoronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in methanol.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
402-66-4 | |
141.10 | |
BXZSBDDOYIWMGC-UHFFFAOYSA-N | |
120232 | |
OC(=O)C1=CN=CC(F)=C1 |
C6H4FNO2 | |
MFCD01318537 | |
5-fluoronicotinic acid, 5-fluoro-3-pyridinecarboxylic acid, 5-fluoro-nicotinic acid, 3-pyridinecarboxylic acid, 5-fluoro, nicotinic acid, 5-fluoro, 3-fluoro-5-carboxypyridine, 3-fluoro-5-pyridinecarboxylic acid, 5-fluoronicotinicacid, pubchem10942 | |
5-fluoropyridine-3-carboxylic acid |
Specifications
402-66-4 | |
White | |
MFCD01318537 | |
5-fluoronicotinic acid, 5-fluoro-3-pyridinecarboxylic acid, 5-fluoro-nicotinic acid, 3-pyridinecarboxylic acid, 5-fluoro, nicotinic acid, 5-fluoro, 3-fluoro-5-carboxypyridine, 3-fluoro-5-pyridinecarboxylic acid, 5-fluoronicotinicacid, pubchem10942 | |
BXZSBDDOYIWMGC-UHFFFAOYSA-N | |
5-fluoropyridine-3-carboxylic acid | |
120232 | |
98% | |
5-Fluoronicotinic acid |
193°C to 198°C | |
C6H4FNO2 | |
5 g | |
Soluble in methanol. | |
OC(=O)C1=CN=CC(F)=C1 | |
141.10 | |
141.1 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : US5745250
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only