missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoronicotinic acid, 98%

Manufacturer:  Alfa Aesar™ H6116214

 View more versions of this product

Catalog No. AAH6116214



Description

Description

Applications
5-Fluoronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

5-Fluoronicotinic acid
193°C to 198°C
402-66-4
MFCD01318537
BXZSBDDOYIWMGC-UHFFFAOYSA-N
5-fluoropyridine-3-carboxylic acid
120232
Powder
White
25g
C6H4FNO2
5-fluoronicotinic acid, 5-fluoro-3-pyridinecarboxylic acid, 5-fluoro-nicotinic acid, 3-pyridinecarboxylic acid, 5-fluoro, nicotinic acid, 5-fluoro, 3-fluoro-5-carboxypyridine, 3-fluoro-5-pyridinecarboxylic acid, 5-fluoronicotinic acid, 5-fluoronicotinicacid, pubchem10942
C1=C(C=NC=C1F)C(=O)O
141.101
141.1
98%

Chemical Identifiers

402-66-4
141.101
BXZSBDDOYIWMGC-UHFFFAOYSA-N
120232
C1=C(C=NC=C1F)C(=O)O
C6H4FNO2
MFCD01318537
5-fluoronicotinic acid, 5-fluoro-3-pyridinecarboxylic acid, 5-fluoro-nicotinic acid, 3-pyridinecarboxylic acid, 5-fluoro, nicotinic acid, 5-fluoro, 3-fluoro-5-carboxypyridine, 3-fluoro-5-pyridinecarboxylic acid, 5-fluoronicotinic acid, 5-fluoronicotinicacid, pubchem10942
5-fluoropyridine-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : US5745250

Documents