Please login to your online account to display your discounted pricing

5-Fluoroindole, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC311920250

 View more versions of this product

Catalog No. AC311920250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 135.14
Quantity 25g
Color Off-white
EINECS Number 206-917-4
Melting Point 44° to 48°C
CAS Max % 100.0
Molecular Formula C8H6FN
CAS Min % 97.5
Packaging Glass bottle
Chemical Name or Material 5-Fluoroindole
Name Note 98%
Assay Percent Range 98%
MDL Number MFCD00005671
CAS 399-52-0
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 5-Fluoroindole
Name Note 98%
CAS 399-52-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H6FN
Formula Weight 135.14
MDL Number MFCD00005671