Please login to your online account to display your discounted pricing

5-Fluoroindole-3-carboxaldehyde, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Fluoroindole-3-carboxaldehyde
Assay Percent Range 99%
Molecular Formula C9H6FNO
Formula Weight 163.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC443350010

  SDS
thermo fisher scientific
AC443350010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC443350050

  SDS
thermo fisher scientific
AC443350050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 163.15
Quantity 1g
Assay Percent Range 99%
Chemical Name or Material 5-Fluoroindole-3-carboxaldehyde
Molecular Formula C9H6FNO
CAS 2338-71-8