Please login to your online account to display your discounted pricing

5'-Fluoroindirubinoxime, Tocris™

FLT3 inhibitor; displays antiproliferative activity

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material 5'-Fluoroindirubinoxime
Assay Percent Range >99%
Molecular Formula C16H10FN3O2
Formula Weight 295.27
Synonym 5'-Fluoro-1H,1'H-[2,3']biindolylidene-3,2'dione 3-oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

403310

 
r & d systems
RDS403310
10mg Each for $225.77

403350

 
r & d systems
RDS403350
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 295.27
Quantity 10mg
Chemical Name or Material 5'-Fluoroindirubinoxime
Assay Percent Range >99%
Molecular Formula C16H10FN3O2
CAS 861214-33-7
Synonym 5'-Fluoro-1H,1'H-[2,3']biindolylidene-3,2'dione 3-oxime
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C