missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Fluorocytosine, TCI America™

Manufacturer:  TCI America F032125G

 View more versions of this product

Catalog No. F032125G



Specifications

Specifications

5-Fluorocytosine
25g
C4H4FN3O
5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
C1=NC(=O)NC(=C1F)N
129.094
CHEBI:5100
Crystal-Powder at 20°C
White
2022-85-7
MFCD00006035
XRECTZIEBJDKEO-UHFFFAOYSA-N
6-amino-5-fluoro-1H-pyrimidin-2-one
3366
129.09
≥98.0% (HPLC,T)

Chemical Identifiers

2022-85-7
129.094
XRECTZIEBJDKEO-UHFFFAOYSA-N
3366
6-amino-5-fluoro-1H-pyrimidin-2-one
C4H4FN3O
MFCD00006035
5-fluorocytosine, flucytosine, ancotil, ancobon, flucytosin, 5-fluorocystosine, fluocytosine, 5-fc, 5-fluorocytosin, fluorcytosine
CHEBI:5100
C1=NC(=O)NC(=C1F)N
Safety and Handling

Safety and Handling

RTECSNumber : HA6040000

TSCA : No

Documents