Please login to your online account to display your discounted pricing

5-Fluoro-DL-tryptophan, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Fluoro-DL-tryptophan
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C11H11FN2O2
Formula Weight 222.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC119680010

  SDS
fisher scientific acros
119680010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Powder
Formula Weight 222.22
Quantity 1g
Color Light beige
Chemical Name or Material 5-Fluoro-DL-tryptophan
Assay Percent Range 99+%
Molecular Formula C11H11FN2O2
CAS 154-08-5
Infrared Spectrum Authentic
Packaging Glass bottle