missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-3-(3-oxycyclopentyl)indole, 95%

$200.28 - $477.72

Chemical Identifiers

CAS 675831-63-7
Molecular Formula C13H12FNO
Molecular Weight (g/mol) 217.243
InChI Key DINNCXVUOXLHSH-UHFFFAOYSA-N
Synonym 3-5-fluoro-1h-indol-3-yl cyclopentan-1-one, 3-5-fluoro-1h-indol-3-yl cyclopentanone, 3-5-fluoro-1h-indol-3-yl-cyclopentanone, 3-5fluoro-1h-indol-3-yl-cyclopentanone
PubChem CID 22139466
IUPAC Name 3-(5-fluoro-1H-indol-3-yl)cyclopentan-1-one
SMILES C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63936MD
View Documents
Alfa Aesar™
H63936MD
250mg Each for $200.28
 
AAH6393603
View Documents
Alfa Aesar™
H6393603
1g Each for $477.72
 
Specifications

Chemical Identifiers

675831-63-7
217.243
3-5-fluoro-1h-indol-3-yl cyclopentan-1-one, 3-5-fluoro-1h-indol-3-yl cyclopentanone, 3-5-fluoro-1h-indol-3-yl-cyclopentanone, 3-5fluoro-1h-indol-3-yl-cyclopentanone
3-(5-fluoro-1H-indol-3-yl)cyclopentan-1-one
C13H12FNO
DINNCXVUOXLHSH-UHFFFAOYSA-N
22139466
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)F

Specifications

250mg
C13H12FNO
DINNCXVUOXLHSH-UHFFFAOYSA-N
3-(5-fluoro-1H-indol-3-yl)cyclopentan-1-one
22139466
Solid
5-Fluoro-3-(3-oxycyclopentyl)indole
675831-63-7
3-5-fluoro-1h-indol-3-yl cyclopentan-1-one, 3-5-fluoro-1h-indol-3-yl cyclopentanone, 3-5-fluoro-1h-indol-3-yl-cyclopentanone, 3-5fluoro-1h-indol-3-yl-cyclopentanone
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)F
217.243
217.24
95%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures