missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-3-(3-oxycyclopentyl)indole, 95%

Manufacturer:  Alfa Aesar™ H63936MD

 View more versions of this product

Catalog No. AAH63936MD



Specifications

Specifications

5-Fluoro-3-(3-oxycyclopentyl)indole
675831-63-7
3-5-fluoro-1h-indol-3-yl cyclopentan-1-one, 3-5-fluoro-1h-indol-3-yl cyclopentanone, 3-5-fluoro-1h-indol-3-yl-cyclopentanone, 3-5fluoro-1h-indol-3-yl-cyclopentanone
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)F
217.243
217.24
95%
250mg
C13H12FNO
DINNCXVUOXLHSH-UHFFFAOYSA-N
3-(5-fluoro-1H-indol-3-yl)cyclopentan-1-one
22139466
Solid

Chemical Identifiers

675831-63-7
217.243
3-5-fluoro-1h-indol-3-yl cyclopentan-1-one, 3-5-fluoro-1h-indol-3-yl cyclopentanone, 3-5-fluoro-1h-indol-3-yl-cyclopentanone, 3-5fluoro-1h-indol-3-yl-cyclopentanone
3-(5-fluoro-1H-indol-3-yl)cyclopentan-1-one
C13H12FNO
DINNCXVUOXLHSH-UHFFFAOYSA-N
22139466
C1CC(=O)CC1C2=CNC3=C2C=C(C=C3)F
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents