Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-3-(1-piperazinylmethyl)Indole, 95%

Chemical Identifiers

Chemical Name or Material 5-Fluoro-3-(1-piperazinylmethyl)Indole
CAS 147595-45-7
Formula Weight 233.28
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6364503

 
alfa aesar™
H6364503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63645MD

 
alfa aesar™
H63645MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6364506

 
alfa aesar™
H6364506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.28
Chemical Name or Material 5-Fluoro-3-(1-piperazinylmethyl)Indole
Physical Form Solid
Assay 95%
CAS 147595-45-7
Quantity 1g