Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-(trifluoromethyl)phenylacetic acid, 97%

$144.80 - $436.96

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)phenylacetic acid
CAS 239135-52-5
Molecular Formula C9H6F4O2
Formula Weight 222.14
MDL Number MFCD00236304
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2669603

  SDS
alfa aesar™
H2669603
1g Each for $144.80

AAH2669606

  SDS
alfa aesar™
H2669606
5g Each for $436.96
Description & Specifications

Specifications

Formula Weight 222.14
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)phenylacetic acid
Assay 97%
Molecular Formula C9H6F4O2
MDL Number MFCD00236304
CAS 239135-52-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.