Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Fluoro-2-(trifluoromethyl)benzyl alcohol, 97%

$67.66 - $296.84

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)benzyl alcohol
CAS 238742-82-0
Molecular Formula C8H6F4O
Formula Weight 194.13
MDL Number MFCD00236286
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2018403

  SDS
alfa aesar™
B2018403
1g Each for $67.66

AAB2018406

  SDS
alfa aesar™
B2018406
5g Each for $296.84
Description & Specifications

Specifications

Formula Weight 194.13
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Quantity 1g
Molecular Formula C8H6F4O
Health Hazard 1 Warning
MDL Number MFCD00236286
Melting Point 48° to 52°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)benzyl alcohol
CAS 238742-82-0
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.